Product Details

SNP ID
rs199915302
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:151518625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGTGCAGAGATGGGCACTCTA[C/T]GTCGAGGTGGACGACGCCCAGCTAA
Phenotype
MIM: 609473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CGN PubMed Links

Gene Details

Gene
CGN
Gene Name
cingulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020770.2 250 Missense Mutation CGT,TGT R36C NP_065821.1
XM_005245365.4 250 Missense Mutation CGT,TGT R36C XP_005245422.1

View Full Product Details