Product Details
- SNP ID
-
rs199967878
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:19666021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGGCCATGCTGAACGCGCTGTAC[C/G]GGCGCTGGGTGCTGGCGCGCGGCCT
- Phenotype
-
MIM: 601109
MIM: 600613
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HTR6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1805054] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HTR6
- Gene Name
- 5-hydroxytryptamine receptor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000871.2 |
735 |
Missense Mutation |
CGG,GGG |
R90G |
NP_000862.1 |
- Gene
- MINOS1-NBL1
- Gene Name
- MINOS1-NBL1 readthrough
There are no transcripts associated with this gene.
- Gene
- NBL1
- Gene Name
- neuroblastoma 1, DAN family BMP antagonist
There are no transcripts associated with this gene.
View Full Product Details