Product Details

SNP ID
rs199861377
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:235450236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTGACATCGACAAGGATCACC[A/G]CACCGTTCCTTCAGTTTCCACAGTT
Phenotype
MIM: 610194 MIM: 604934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GALNT2 PubMed Links

Gene Details

Gene
B3GALNT2
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277155.2 1789 Intron NP_001264084.1
NM_152490.4 1789 Silent Mutation TGC,TGT C491C NP_689703.1
XM_006711749.3 1789 Intron XP_006711812.1
XM_017000394.1 1789 Silent Mutation TGC,TGT C532C XP_016855883.1
XM_017000395.1 1789 UTR 3 XP_016855884.1
Gene
TBCE
Gene Name
tubulin folding cofactor E
There are no transcripts associated with this gene.

View Full Product Details