Product Details

SNP ID
rs200750484
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:74198268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTGTGCGGCGCGGTCTCAGGG[A/T]AGGTGGGGCTATGGCAGCTGCTAGG
Phenotype
MIM: 603609
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FPGT PubMed Links

Gene Details

Gene
FPGT
Gene Name
fucose-1-phosphate guanylyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199328.2 57 Missense Mutation GAA,GTA E10V NP_001186257.2
NM_001199329.2 57 Missense Mutation GAA,GTA E10V NP_001186258.2
NM_003838.4 57 Missense Mutation GAA,GTA E10V NP_003829.3
Gene
FPGT-TNNI3K
Gene Name
FPGT-TNNI3K readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112808.2 57 Missense Mutation GAA,GTA E10V NP_001106279.2
NM_001199327.1 57 Missense Mutation GAA,GTA E10V NP_001186256.2
Gene
LRRIQ3
Gene Name
leucine rich repeats and IQ motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105659.1 57 Intron NP_001099129.1
NM_001322315.1 57 Intron NP_001309244.1
XM_011540658.1 57 Intron XP_011538960.1
XM_011540659.1 57 Intron XP_011538961.1
XM_011540660.1 57 Intron XP_011538962.1
XM_011540662.2 57 Intron XP_011538964.1
XM_011540663.2 57 Intron XP_011538965.1
XM_017000286.1 57 Intron XP_016855775.1
XM_017000287.1 57 Intron XP_016855776.1
XM_017000288.1 57 Intron XP_016855777.1

View Full Product Details