Product Details
- SNP ID
-
rs200460052
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:232805421 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTTCAGCTTCTCGTTTGCGGAGC[C/G]CGCGGCGGCGTTTCCTGGGGCAACA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MAP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12409898] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MAP10
- Gene Name
- microtubule associated protein 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019090.2 |
530 |
Missense Mutation |
CCC,CGC |
P133R |
NP_061963.2 |
View Full Product Details