Product Details

SNP ID
rs200208332
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:192810395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCTCCTGAGGAAGCACAGCTG[C/T]GGTCAGAAGCATTTGACGAGCTGCT
Phenotype
MIM: 600861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS2 PubMed Links

Gene Details

Gene
RGS2
Gene Name
regulator of G-protein signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002923.3 271 Missense Mutation CGG,TGG R80W NP_002914.1

View Full Product Details