Product Details
- SNP ID
-
rs200620700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:154228669 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATGTGTGATTGCTTTGCATGAC[G/T]GCAATGGAGATGTCAACAGAGCTAT
- Phenotype
-
MIM: 616472
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf43
PubMed Links
Gene Details
- Gene
- C1orf43
- Gene Name
- chromosome 1 open reading frame 43
There are no transcripts associated with this gene.
- Gene
- UBAP2L
- Gene Name
- ubiquitin associated protein 2 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127320.2 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
NP_001120792.1 |
NM_001287815.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
NP_001274744.1 |
NM_001287816.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
NP_001274745.1 |
NM_014847.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
NP_055662.3 |
XM_005245658.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245715.1 |
XM_005245667.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245724.1 |
XM_005245668.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245725.1 |
XM_005245669.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245726.1 |
XM_005245670.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245727.1 |
XM_005245672.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245729.1 |
XM_005245673.3 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245730.1 |
XM_005245674.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_005245731.1 |
XM_011510215.2 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_011508517.1 |
XM_011510216.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_011508518.1 |
XM_017002972.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858461.1 |
XM_017002973.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858462.1 |
XM_017002974.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858463.1 |
XM_017002975.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858464.1 |
XM_017002976.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858465.1 |
XM_017002977.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858466.1 |
XM_017002978.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858467.1 |
XM_017002979.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858468.1 |
XM_017002980.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858469.1 |
XM_017002981.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858470.1 |
XM_017002982.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858471.1 |
XM_017002983.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858472.1 |
XM_017002984.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858473.1 |
XM_017002985.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858474.1 |
XM_017002986.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858475.1 |
XM_017002987.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858476.1 |
XM_017002988.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858477.1 |
XM_017002989.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858478.1 |
XM_017002990.1 |
390 |
Missense Mutation |
GGC,TGC |
G75C |
XP_016858479.1 |
XM_017002991.1 |
390 |
UTR 5 |
|
|
XP_016858480.1 |
View Full Product Details