Product Details

SNP ID
rs200462252
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:78628096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGCTTACATTGATTACAATATG[A/G]TTGTAGCCTTTATGCTTGGAAATTA
Phenotype
MIM: 613975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI44L PubMed Links

Gene Details

Gene
IFI44L
Gene Name
interferon induced protein 44 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006820.3 219 Missense Mutation ATT,GTT I61V NP_006811.2
XM_005270391.3 219 Intron XP_005270448.1
XM_005270392.4 219 Intron XP_005270449.1
XM_005270393.4 219 Intron XP_005270450.1
XM_006710304.3 219 Intron XP_006710367.1
XM_011540539.2 219 Missense Mutation ATT,GTT I61V XP_011538841.1
XM_017000120.1 219 Intron XP_016855609.1

View Full Product Details