Product Details

SNP ID
rs200369681
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:203863668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTTATGATTATTTCAGAGATC[A/G]CGGATTCAGGTGTGGCTCTATGAGC
Phenotype
MIM: 128260 MIM: 613513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNRPE PubMed Links

Gene Details

Gene
SNRPE
Gene Name
small nuclear ribonucleoprotein polypeptide E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304464.1 162 Silent Mutation TCA,TCG S21S NP_001291393.1
NM_003094.3 162 Silent Mutation TCA,TCG S29S NP_003085.1
Gene
ZC3H11A
Gene Name
zinc finger CCCH-type containing 11A
There are no transcripts associated with this gene.

View Full Product Details