Product Details

SNP ID
rs200463902
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:228408666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCTGCAGAACCCACTGCCCTC[C/T]GGCGAAGAGCCGAGGTAGCGCCTCT
Phenotype
MIM: 607868 MIM: 606123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM11 PubMed Links

Gene Details

Gene
TRIM11
Gene Name
tripartite motif containing 11
There are no transcripts associated with this gene.

Gene
TRIM17
Gene Name
tripartite motif containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024940.2 1886 Silent Mutation CCA,CCG P323P NP_001020111.1
NM_001134855.1 1886 Intron NP_001128327.1
NM_016102.3 1886 Silent Mutation CCA,CCG P323P NP_057186.1
XM_006711779.2 1886 Silent Mutation CCA,CCG P323P XP_006711842.1
XM_011544209.2 1886 Silent Mutation CCA,CCG P323P XP_011542511.1
XM_011544210.2 1886 Silent Mutation CCA,CCG P323P XP_011542512.1
XM_011544211.2 1886 Silent Mutation CCA,CCG P296P XP_011542513.1
XM_017001418.1 1886 UTR 3 XP_016856907.1
XM_017001419.1 1886 UTR 3 XP_016856908.1

View Full Product Details