Product Details
- SNP ID
-
rs200661833
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:43949419 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGTCCTGTGCTGTCCTGCAGGC[A/G]CTGCACCAGCTCTACTATGATCCCA
- Phenotype
-
MIM: 610411
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IPO13
PubMed Links
Gene Details
- Gene
- IPO13
- Gene Name
- importin 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014652.3 |
882 |
Silent Mutation |
GCA,GCG |
A29A |
NP_055467.3 |
View Full Product Details