Product Details
- SNP ID
-
rs200623969
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:5865178 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCCCCGAAGGACAAGGGACAGG[C/T]GGGTCAGCTGGCCTGCGACGCAGGA
- Phenotype
-
MIM: 607215
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4689
PubMed Links
Gene Details
- Gene
- MIR4689
- Gene Name
- microRNA 4689
There are no transcripts associated with this gene.
- Gene
- NPHP4
- Gene Name
- nephronophthisis 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001291593.1 |
3045 |
Missense Mutation |
CAC,CGC |
H734R |
NP_001278522.1 |
NM_001291594.1 |
3045 |
Missense Mutation |
CAC,CGC |
H735R |
NP_001278523.1 |
NM_015102.4 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
NP_055917.1 |
XM_006710563.3 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
XP_006710626.1 |
XM_011541213.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1246R |
XP_011539515.1 |
XM_011541214.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1233R |
XP_011539516.1 |
XM_011541215.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1210R |
XP_011539517.1 |
XM_011541216.2 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
XP_011539518.1 |
XM_011541217.2 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
XP_011539519.1 |
XM_011541218.2 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
XP_011539520.1 |
XM_017000996.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1232R |
XP_016856485.1 |
XM_017000997.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1247R |
XP_016856486.1 |
XM_017000998.1 |
3045 |
Intron |
|
|
XP_016856487.1 |
XM_017000999.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1071R |
XP_016856488.1 |
XM_017001000.1 |
3045 |
Missense Mutation |
CAC,CGC |
H1071R |
XP_016856489.1 |
XM_017001001.1 |
3045 |
Missense Mutation |
CAC,CGC |
H981R |
XP_016856490.1 |
XM_017001002.1 |
3045 |
Intron |
|
|
XP_016856491.1 |
XM_017001003.1 |
3045 |
Missense Mutation |
CAC,CGC |
H734R |
XP_016856492.1 |
View Full Product Details