Product Details

SNP ID
rs200225722
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:37806886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTCAGAGAACTGGCCTGGGAGC[C/T]GAGGTTGGCACTAGTGAGAGCAAGC
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links
Additional Information
For this assay, SNP(s) [rs2291297] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142726.1 608 Intron NP_001136198.1
NM_198446.2 608 Intron NP_940848.2
Gene
MANEAL
Gene Name
mannosidase endo-alpha like
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024640.3 608 Missense Mutation AGC,GGC S199G NP_078916.3

View Full Product Details