Product Details

SNP ID
rs200582860
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:247100564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCGTGGAGGTGAAGGGAACTG[A/G]GGCGACTGAAGGCTTGATCACATTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf229 PubMed Links

Gene Details

Gene
C1orf229
Gene Name
chromosome 1 open reading frame 229
There are no transcripts associated with this gene.

Gene
ZNF669
Gene Name
zinc finger protein 669
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142572.1 1087 Missense Mutation CCC,CTC P316L NP_001136044.1
NM_024804.2 1087 Missense Mutation CCC,CTC P402L NP_079080.2
XM_005273270.2 1087 Missense Mutation CCC,CTC P279L XP_005273327.1
XM_005273271.2 1087 Missense Mutation CCC,CTC P279L XP_005273328.1

View Full Product Details