Product Details

SNP ID
rs200663569
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:156061418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCAAGATGGGGAATGGAACTGA[A/G]GAAGATTATAACTTTGTCTTCAAGG
Phenotype
MIM: 610389 MIM: 612942 MIM: 605440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
There are no transcripts associated with this gene.

Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020387.3 270 Silent Mutation GAA,GAG E6E NP_065120.2
Gene
UBQLN4
Gene Name
ubiquilin 4
There are no transcripts associated with this gene.

View Full Product Details