Product Details

SNP ID
rs200770175
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:153149928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATACTTCTGCTGGCAGGGTGG[G/T]TATGGTTGCACAGGAGGGCATTTAT
Phenotype
MIM: 182269
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPRR2C PubMed Links

Gene Details

Gene
SPRR2C
Gene Name
small proline rich protein 2C (pseudogene)
There are no transcripts associated with this gene.

Gene
SPRR2G
Gene Name
small proline rich protein 2G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014291.3 227 Nonsense Mutation TAA,TAC *61Y NP_001014313.1
XM_017002177.1 227 Nonsense Mutation TAA,TAC *61Y XP_016857666.1
XM_017002178.1 227 Nonsense Mutation TAA,TAC *61Y XP_016857667.1

View Full Product Details