Product Details

SNP ID
rs200809621
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:52020991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCACCCGTCAGCCTTTCCTGA[A/G]CTTCCTTCATCCCCTGAACAACTGT
Phenotype
MIM: 609448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNDC12 PubMed Links

Gene Details

Gene
TXNDC12
Gene Name
thioredoxin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015913.3 1536 Missense Mutation GCT,GTT A154V NP_056997.1

View Full Product Details