Product Details

SNP ID
rs200319146
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:155760620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGTCTTCATACTTGCCAGGGA[A/T]ATGTTGTAGGGCTTCTCGCACCTAC
Phenotype
MIM: 610393
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GON4L PubMed Links

Gene Details

Gene
GON4L
Gene Name
gon-4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282856.1 5072 Missense Mutation ATC,TTC I1645F NP_001269785.1
NM_001282858.1 5072 Missense Mutation ATC,TTC I1645F NP_001269787.1
NM_001282860.1 5072 Missense Mutation ATC,TTC I1645F NP_001269789.1
NM_001282861.1 5072 Intron NP_001269790.1
NM_032292.5 5072 Intron NP_115668.4
XM_005245284.3 5072 Missense Mutation ATC,TTC I1473F XP_005245341.1
XM_005245286.3 5072 Missense Mutation ATC,TTC I841F XP_005245343.1
XM_006711393.3 5072 Missense Mutation ATC,TTC I1645F XP_006711456.1
XM_006711394.3 5072 Missense Mutation ATC,TTC I1645F XP_006711457.1
XM_011509658.2 5072 Missense Mutation ATC,TTC I1616F XP_011507960.1
XM_011509659.2 5072 Missense Mutation ATC,TTC I1473F XP_011507961.1
Gene
MSTO2P
Gene Name
misato family member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details