Product Details

SNP ID
rs200147905
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1326991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCTCGATGAGAAGGAAGAGGT[A/C]TTGCTGGACCCCTACATTGCCAGCT
Phenotype
MIM: 611354 MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPSF3L PubMed Links

Gene Details

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029885.1 496 Silent Mutation GTA,GTC V27V NP_001025056.1
XM_005244801.3 496 Silent Mutation GTA,GTC V27V XP_005244858.1
XM_005244802.1 496 Intron XP_005244859.1
XM_011542200.2 496 Silent Mutation GTA,GTC V27V XP_011540502.1
Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
There are no transcripts associated with this gene.

View Full Product Details