Product Details

SNP ID
rs200254500
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:172659272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCAGTGCCAGCTCTCCCTGGG[A/C]CCCTCCAGGCACAGTTCTTCCCTGT
Phenotype
MIM: 134638
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FASLG PubMed Links

Gene Details

Gene
FASLG
Gene Name
Fas ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000639.2 265 Missense Mutation GAC,GCC D24A NP_000630.1
NM_001302746.1 265 Missense Mutation GAC,GCC D24A NP_001289675.1

View Full Product Details