Product Details

SNP ID
rs200333134
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:32209102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTGAGGATACACTATGGCAGG[G/T]GGCAGTCCAGCAGCCAAGAGGGTAG
Phenotype
MIM: 610162
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC28B PubMed Links

Gene Details

Gene
CCDC28B
Gene Name
coiled-coil domain containing 28B
There are no transcripts associated with this gene.

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099434.1 290 Silent Mutation GGG,GGT G3G NP_001092904.1
XM_017000413.1 290 Silent Mutation GGG,GGT G3G XP_016855902.1
XM_017000414.1 290 Intron XP_016855903.1
Gene
IQCC
Gene Name
IQ motif containing C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160042.1 290 Intron NP_001153514.1
NM_018134.2 290 Intron NP_060604.2
Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 290 Intron NP_061991.3
XM_011541795.2 290 Intron XP_011540097.1
XM_017001812.1 290 Intron XP_016857301.1
XM_017001813.1 290 Intron XP_016857302.1
XM_017001814.1 290 Intron XP_016857303.1
XM_017001815.1 290 Intron XP_016857304.1
XM_017001816.1 290 Intron XP_016857305.1
XM_017001817.1 290 Intron XP_016857306.1
XM_017001818.1 290 Intron XP_016857307.1
XM_017001819.1 290 Intron XP_016857308.1
XM_017001820.1 290 Intron XP_016857309.1
XM_017001821.1 290 Intron XP_016857310.1
XM_017001822.1 290 Intron XP_016857311.1
XM_017001823.1 290 Intron XP_016857312.1
XM_017001824.1 290 Intron XP_016857313.1
XM_017001825.1 290 Intron XP_016857314.1
XM_017001826.1 290 Intron XP_016857315.1
XM_017001827.1 290 Intron XP_016857316.1
XM_017001828.1 290 Intron XP_016857317.1
XM_017001829.1 290 Intron XP_016857318.1
XM_017001830.1 290 Intron XP_016857319.1
XM_017001831.1 290 Intron XP_016857320.1

View Full Product Details