Product Details
- SNP ID
-
rs200373449
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:19074873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGGTCCTTCAGGAAGCTCTCTG[C/G]ATCGGTGATTTCTGATAAAAGACCT
- Phenotype
-
MIM: 609890
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
UBR4
PubMed Links
Gene Details
- Gene
- UBR4
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020765.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5171A |
NP_065816.2 |
XM_011541108.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5222A |
XP_011539410.1 |
XM_011541109.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5221A |
XP_011539411.1 |
XM_011541110.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5221A |
XP_011539412.1 |
XM_011541111.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5221A |
XP_011539413.1 |
XM_011541112.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5217A |
XP_011539414.1 |
XM_011541113.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5216A |
XP_011539415.1 |
XM_011541114.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5216A |
XP_011539416.1 |
XM_011541115.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5214A |
XP_011539417.1 |
XM_011541116.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5211A |
XP_011539418.1 |
XM_011541117.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5194A |
XP_011539419.1 |
XM_011541118.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5193A |
XP_011539420.1 |
XM_011541119.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5182A |
XP_011539421.1 |
XM_011541120.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5181A |
XP_011539422.1 |
XM_011541121.2 |
15492 |
Missense Mutation |
CCA,GCA |
P5170A |
XP_011539423.1 |
XM_017000822.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5215A |
XP_016856311.1 |
XM_017000823.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5206A |
XP_016856312.1 |
XM_017000824.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5188A |
XP_016856313.1 |
XM_017000825.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5183A |
XP_016856314.1 |
XM_017000826.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5182A |
XP_016856315.1 |
XM_017000827.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5177A |
XP_016856316.1 |
XM_017000828.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5169A |
XP_016856317.1 |
XM_017000829.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5153A |
XP_016856318.1 |
XM_017000830.1 |
15492 |
Missense Mutation |
CCA,GCA |
P5136A |
XP_016856319.1 |
View Full Product Details