Product Details

SNP ID
rs200215550
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:248387654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACCAGAGGCTTTACCCTCATG[G/T]GGCTCTTCACTCACAATAAATGCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2T6 PubMed Links

Gene Details

Gene
OR2T6
Gene Name
olfactory receptor family 2 subfamily T member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005471.1 Intron NP_001005471.1

View Full Product Details