Product Details

SNP ID
rs200294153
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26950113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTGCCCCCGACGAGTCTGTGT[C/T]GGTGCCCTGGAAGGATGAGTCATGG
Phenotype
MIM: 616758 MIM: 610325
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDF1 PubMed Links

Gene Details

Gene
KDF1
Gene Name
keratinocyte differentiation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152365.2 1246 Missense Mutation AAC,GAC N385D NP_689578.2
XM_005245735.2 1246 Missense Mutation AAC,GAC N385D XP_005245792.1
XM_011540622.1 1246 Missense Mutation AAC,GAC N385D XP_011538924.1
Gene
NUDC
Gene Name
nuclear distribution C, dynein complex regulator
There are no transcripts associated with this gene.

View Full Product Details