Product Details

SNP ID
rs200613814
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:175323397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGTTTTCTCCCTGCCATGAGA[C/T]GGTGGTTGTAGGGGCGCATCTTCAT
Phenotype
MIM: 601995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNR PubMed Links

Gene Details

Gene
TNR
Gene Name
tenascin R
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003285.2 4201 Missense Mutation CAT,CGT H1346R NP_003276.3
XM_011509949.2 4201 Missense Mutation CAT,CGT H1187R XP_011508251.1
XM_017002218.1 4201 Missense Mutation CAT,CGT H1346R XP_016857707.1
XM_017002219.1 4201 Missense Mutation CAT,CGT H1346R XP_016857708.1

View Full Product Details