Product Details
- SNP ID
-
rs200242493
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:152515434 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGCCAAAGGCTTTTCCAAGGGGTC[G/T]TCCCAGGGCCCCGCTCCGTGTCCCG
- Phenotype
-
MIM: 612619
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRCT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73004856] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRCT1
- Gene Name
- cysteine rich C-terminal 1
- Gene
- LCE5A
- Gene Name
- late cornified envelope 5A
There are no transcripts associated with this gene.
View Full Product Details