Product Details

SNP ID
rs200667184
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:160370883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGCCCGAGGCCCAGAGCCGG[C/G]AGAGCCTGGCCGGAAAGACCTGCAG
Phenotype
MIM: 162360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NHLH1 PubMed Links

Gene Details

Gene
NHLH1
Gene Name
nescient helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005598.3 594 Missense Mutation GCA,GGA A51G NP_005589.1

View Full Product Details