Product Details
- SNP ID
-
rs200138400
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:247815241 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATGTTGGACCCACAGTAGGATAA[G/T]GAGAAGGTGCCAGCTGTGTGCATCA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR14A16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35979231] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR14A16
- Gene Name
- olfactory receptor family 14 subfamily A member 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001966.1 |
489 |
Silent Mutation |
TCA,TCC |
S163S |
NP_001001966.1 |
View Full Product Details