Product Details

SNP ID
rs200587705
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:147908311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGGAGCTGGGCCAGCAGGCGG[A/G]GACTAACGGCGGCCCGGACCAGGGC
Phenotype
MIM: 600897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJA8 PubMed Links

Gene Details

Gene
GJA8
Gene Name
gap junction protein alpha 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005267.4 1156 Missense Mutation GAG,GGG E119G NP_005258.2
XM_011509416.1 1156 Missense Mutation GAG,GGG E119G XP_011507718.1
XM_011509417.1 1156 Missense Mutation GAG,GGG E119G XP_011507719.1

View Full Product Details