Product Details

SNP ID
rs200668040
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156058024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTGGCCAACCTTCTGCTGTGTA[C/T]GTATGCCAAGGAGACCGTGGGCTTT
Phenotype
MIM: 610389 MIM: 612942 MIM: 605440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145264.1 442 Intron NP_001138736.1
NM_014017.3 442 Missense Mutation ACG,ATG T93M NP_054736.1
Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
UBQLN4
Gene Name
ubiquilin 4
There are no transcripts associated with this gene.

View Full Product Details