Product Details

SNP ID
rs200801783
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:108648840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGGGCCTTTGAGCCACCAATG[C/T]CTTTGGGCTTATCACCCCGTTCCCC
Phenotype
MIM: 612178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
There are no transcripts associated with this gene.

Gene
HENMT1
Gene Name
HEN1 methyltransferase homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102592.1 1329 Missense Mutation GAC,GGC D303G NP_001096062.1
NM_144584.2 1329 Missense Mutation GAC,GGC D303G NP_653185.2
XM_005270411.1 1329 Missense Mutation GAC,GGC D311G XP_005270468.1
XM_017000179.1 1329 Missense Mutation GAC,GGC D147G XP_016855668.1

View Full Product Details