Product Details

SNP ID
rs200205424
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:158606486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGGGATTTTGTCTTCCTGGG[C/T]TTCTCCAGTTCTGGGGAGTTGCAGC
Phenotype
MIM: 182860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10Z1 PubMed Links

Gene Details

Gene
OR10Z1
Gene Name
olfactory receptor family 10 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004478.1 48 Silent Mutation GGC,GGT G16G NP_001004478.1
Gene
SPTA1
Gene Name
spectrin alpha, erythrocytic 1
There are no transcripts associated with this gene.

View Full Product Details