Product Details

SNP ID
rs200417619
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:109475596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCTCAGCTCTTTGCTATTTTC[A/G]CCTTCGGGTCCTGTGGCTCCTACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN7L2 PubMed Links

Gene Details

Gene
ATXN7L2
Gene Name
ataxin 7 like 2
There are no transcripts associated with this gene.

Gene
SYPL2
Gene Name
synaptophysin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040709.1 511 Missense Mutation ACC,GCC T49A NP_001035799.1
XM_011541283.2 511 Missense Mutation ACC,GCC T49A XP_011539585.1
XM_011541284.2 511 Missense Mutation ACC,GCC T49A XP_011539586.1
XM_011541285.1 511 Missense Mutation ACC,GCC T49A XP_011539587.1

View Full Product Details