Product Details

SNP ID
rs200895543
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:12644610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTTTTCCTGGGGGTCTTTGTC[G/T]GGGCTGTCTTTGAGCACTTCCTCAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AADACL4 PubMed Links

Gene Details

Gene
AADACL4
Gene Name
arylacetamide deacetylase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013630.1 64 Missense Mutation GGG,TGG G22W NP_001013652.1
XM_017001152.1 64 Intron XP_016856641.1
XM_017001153.1 64 Intron XP_016856642.1

View Full Product Details