Product Details

SNP ID
rs200126811
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:169514452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCCTTTGGTATTAGTATTTCCT[G/T]CAAAAATCTGAAAGCCAAATAAGAG
Phenotype
MIM: 612309
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
F5 PubMed Links

Gene Details

Gene
F5
Gene Name
coagulation factor V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000130.4 6681 Missense Mutation GAA,GCA E2179A NP_000121.2
XM_017000660.1 6681 Missense Mutation GAA,GCA E2042A XP_016856149.1

View Full Product Details