Product Details

SNP ID
rs200643907
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:151801337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGACCCTTACTCTCCCAGCCCT[A/G]CCCAGCCAAGCCCCATGAGGCCTCA
Phenotype
MIM: 609794 MIM: 602943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004432.3 1594 Silent Mutation GGC,GGT G456G NP_001004432.1
Gene
RORC
Gene Name
RAR related orphan receptor C
There are no transcripts associated with this gene.

View Full Product Details