Product Details

SNP ID
rs200670701
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:37815298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGGGTCTGAAGGAGTCTCTGC[C/T]TGTCGGCTTTGCTCACAGGAGGAGG
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005955.2 2405 Silent Mutation CAA,CAG Q700Q NP_005946.2
XM_011541491.2 2405 Silent Mutation CAA,CAG Q700Q XP_011539793.1
XM_011541493.2 2405 Intron XP_011539795.1
XM_011541494.1 2405 Silent Mutation CAA,CAG Q391Q XP_011539796.1
Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
There are no transcripts associated with this gene.

View Full Product Details