Product Details

SNP ID
rs202086339
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:109917360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGACATAATGGAGGACACCATGC[A/G]CTTCAGAGATAACACCCCCAATGCC
Phenotype
MIM: 120420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF1 PubMed Links

Gene Details

Gene
CSF1
Gene Name
colony stimulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000757.5 706 Missense Mutation CAC,CGC H98R NP_000748.3
NM_172210.2 706 Missense Mutation CAC,CGC H98R NP_757349.1
NM_172211.3 706 Missense Mutation CAC,CGC H98R NP_757350.1
NM_172212.2 706 Missense Mutation CAC,CGC H98R NP_757351.1
XM_017000369.1 706 Missense Mutation CAC,CGC H57R XP_016855858.1

View Full Product Details