Product Details

SNP ID
rs202022112
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:155900391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAACACTTCACATCTTCTCTTCA[A/G]GTTACTGAATCTTTCTTCTTCCGGA
Phenotype
MIM: 609591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIT1 PubMed Links

Gene Details

Gene
RIT1
Gene Name
Ras like without CAAX 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256820.1 860 Silent Mutation ACC,ACT T183T NP_001243749.1
NM_001256821.1 860 Silent Mutation ACC,ACT T236T NP_001243750.1
NM_006912.5 860 Silent Mutation ACC,ACT T219T NP_008843.1

View Full Product Details