Product Details

SNP ID
rs201903475
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:151055701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCTGGGGTTTCTCTACCACACA[A/G]CAGCCCAGTTTGTGCCAAAATTCAC
Phenotype
MIM: 611275 MIM: 604684
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BNIPL PubMed Links

Gene Details

Gene
BNIPL
Gene Name
BCL2 interacting protein like
There are no transcripts associated with this gene.

Gene
C1orf56
Gene Name
chromosome 1 open reading frame 56
There are no transcripts associated with this gene.

Gene
CDC42SE1
Gene Name
CDC42 small effector 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038707.1 464 Silent Mutation NP_001033796.1
NM_020239.3 464 Silent Mutation NP_064624.1
XM_017001847.1 464 Silent Mutation XP_016857336.1
Gene
MLLT11
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
There are no transcripts associated with this gene.

View Full Product Details