Product Details

SNP ID
rs202153297
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:54036618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAATTAAATTTACCTCAGAGG[A/G]AACATACTGCTCCACAGCTGTAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 731 Missense Mutation CCC,TCC P271S NP_001291972.1
NM_001305049.1 731 Missense Mutation CCC,TCC P140S NP_001291978.1
NM_001305050.1 731 Missense Mutation CCC,TCC P204S NP_001291979.1
NM_001305051.1 731 Missense Mutation CCC,TCC P139S NP_001291980.1
NM_001305052.1 731 Missense Mutation CCC,TCC P139S NP_001291981.1
NM_001305066.1 731 Intron NP_001291995.1
NM_004872.4 731 Missense Mutation CCC,TCC P270S NP_004863.2
XM_006711051.1 731 Missense Mutation CCC,TCC P203S XP_006711114.1

View Full Product Details