Product Details

SNP ID
rs202221916
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:32470991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGACATACATTAGGTCATCCCTC[C/G]ACATTTGCCGAAAGATGGAGAAGGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB8B PubMed Links

Gene Details

Gene
ZBTB8B
Gene Name
zinc finger and BTB domain containing 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145720.1 457 Missense Mutation CAC,GAC H123D NP_001139192.1

View Full Product Details