Product Details
- SNP ID
-
rs202037434
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:53088037 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACATTGGTCTCCAGCTCACTGATC[C/T]GGATGGTGCAGTGGTTCAGACTCGC
- Phenotype
-
MIM: 608661
MIM: 604471
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PODN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1288400] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PODN
- Gene Name
- podocan
There are no transcripts associated with this gene.
- Gene
- SLC1A7
- Gene Name
- solute carrier family 1 member 7
View Full Product Details