Product Details

SNP ID
rs201853353
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1374433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCACTCCAGAGACGGGCCAA[A/G]GCGGGCGGCCGCCTGCAGAAAACCA
Phenotype
MIM: 609183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKAIP1 PubMed Links

Gene Details

Gene
AURKAIP1
Gene Name
aurora kinase A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127229.1 239 Missense Mutation CCT,CTT P22L NP_001120701.1
NM_001127230.1 239 Missense Mutation CCT,CTT P22L NP_001120702.1
NM_017900.2 239 Missense Mutation CCT,CTT P22L NP_060370.1
XM_011541670.2 239 Missense Mutation CCT,CTT P51L XP_011539972.1

View Full Product Details