Product Details

SNP ID
rs201972571
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:227816184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGTCAGCGTGCACTACGCAGG[A/C]CTCCACGTCTGCGGCGGCTCCATCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRSS38 PubMed Links

Gene Details

Gene
PRSS38
Gene Name
protease, serine 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183062.2 243 Silent Mutation GGA,GGC G81G NP_898885.1
XM_011544175.2 243 Silent Mutation GGA,GGC G81G XP_011542477.1
XM_011544176.2 243 Silent Mutation GGA,GGC G81G XP_011542478.1

View Full Product Details