Product Details

SNP ID
rs201840590
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:86560334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCTGACCTTCTACTTGGAAAA[A/C]AACAAAATGAATATGGACCACCAGG
Phenotype
MIM: 616857
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 468 Missense Mutation AAA,CAA K142Q NP_036260.2
XM_011541015.2 468 Missense Mutation AAA,CAA K91Q XP_011539317.1

View Full Product Details