Product Details

SNP ID
rs202211100
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:37807112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTTGACTCACAGGCGTAAAAG[C/G]GTTTAGGTCCTTGTTGAGCTCCTCC
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142726.1 506 Intron NP_001136198.1
NM_198446.2 506 Intron NP_940848.2
Gene
MANEAL
Gene Name
mannosidase endo-alpha like
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024640.3 506 Missense Mutation CCT,GCT P165A NP_078916.3

View Full Product Details