Product Details

SNP ID
rs201920792
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:169515520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTGTAGTGGATGGTATAGCTC[C/T]TTACATACATTTCAGAGGACAGAGA
Phenotype
MIM: 612309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F5 PubMed Links

Gene Details

Gene
F5
Gene Name
coagulation factor V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000130.4 6597 Missense Mutation AAG,AGG K2151R NP_000121.2
XM_017000660.1 6597 Missense Mutation AAG,AGG K2014R XP_016856149.1

View Full Product Details