Product Details

SNP ID
rs202131118
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:153982426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGACTTGAGCAAGATGTCCC[A/G]GGCCAGGGAACTAAAAGCCTAAAGT
Phenotype
MIM: 607138 MIM: 604671 MIM: 602672 MIM: 603702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
There are no transcripts associated with this gene.

Gene
JTB
Gene Name
jumping translocation breakpoint
There are no transcripts associated with this gene.

Gene
RAB13
Gene Name
RAB13, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272038.1 758 Missense Mutation NP_001258967.1
NM_002870.3 758 Missense Mutation NP_002861.1
XM_017001959.1 758 Intron XP_016857448.1
Gene
RPS27
Gene Name
ribosomal protein S27
There are no transcripts associated with this gene.

View Full Product Details