Product Details

SNP ID
rs202172237
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:223394099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGAGGCACTCAGCCTGCGTGTG[C/T]GCCCAGAAGAGAGACAGCAGCGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC185 PubMed Links

Gene Details

Gene
CCDC185
Gene Name
coiled-coil domain containing 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152610.2 727 Silent Mutation TGC,TGT C208C NP_689823.2

View Full Product Details